Assay Details
| Gene Symbol: ccni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102993.1 | NP_001096463.1 | 2 - 3 | 434 | 69 | |
| GenBank mRNA | BC136194.1 | - | 2 - 3 | 434 | 69 | |
More Information
Set Membership: |
|

ccni2
Genomic Map