Assay Details
| Gene Symbol: ccni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102993.1 | NP_001096463.1 | 5 - 6 | 783 | 82 | |
| XM_018091914.1 | XP_017947403.1 | 5 - 6 | 1108 | 82 | ||
| GenBank mRNA | BC090569.1 | - | 4 - 5 | 592 | 82 | |
| BC136194.1 | - | 5 - 6 | 783 | 82 | ||
More Information
Set Membership: |
|

ccni2
Genomic Map