Assay Details
| Gene Symbol: mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102999.1 | NP_001096469.1 | 6 - 7 | 1316 | 60 | |
| XM_012966783.2 | XP_012822237.1 | 6 - 7 | 1412 | 60 | ||
| GenBank mRNA | BC136206.1 | - | 6 - 7 | 1316 | 60 | |
More Information
Set Membership: |
|

mmp17
Genomic Map