Assay Details
| Gene Symbol: prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103036.1 | NP_001096506.1 | 9 - 10 | 714 | 82 | |
| XM_012956318.2 | XP_012811772.1 | 9 - 10 | 755 | 82 | ||
| GenBank mRNA | BC135379.1 | - | 9 - 10 | 714 | 82 | |
More Information
Set Membership: |
|

prkag1
Genomic Map