Assay Details
| Gene Symbol: ubl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103040.1 | NP_001096510.1 | 2 - 3 | 167 | 78 | |
| GenBank mRNA | BC135438.1 | - | 2 - 3 | 167 | 78 | |
More Information
Set Membership: |
|

ubl4a
Genomic Map