Assay Details
| Gene Symbol: gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103041.1 | NP_001096511.1 | 3 - 4 | 268 | 100 | |
| GenBank mRNA | BC135448.1 | - | 3 - 4 | 268 | 100 | |
More Information
Set Membership: |
|

gabarapl2
Genomic Map