Assay Details
| Gene Symbol: nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103042.1 | NP_001096512.1 | 3 - 4 | 308 | 64 | |
| GenBank mRNA | BC135468.1 | - | 3 - 4 | 308 | 64 | |
More Information
Set Membership: |
|

nip7
Genomic Map