Assay Details
| Gene Symbol: setd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103050.1 | NP_001096520.1 | 7 - 8 | 925 | 73 | |
| XM_012960485.1 | XP_012815939.1 | 7 - 8 | 898 | 73 | ||
| GenBank mRNA | BC135640.1 | - | 7 - 8 | 925 | 73 | |
More Information
Set Membership: |
|

setd6
Genomic Map