Assay Details
| Gene Symbol: tomm70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103057.1 | NP_001096527.1 | 10 - 11 | 1486 | 69 | |
| GenBank mRNA | BC084905.1 | - | 10 - 11 | 1485 | 69 | |
| BC135771.1 | - | 10 - 11 | 1486 | 69 | ||
More Information
Set Membership: |
|

tomm70
Genomic Map