Assay Details
| Gene Symbol: g6pc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103061.1 | NP_001096531.1 | 4 - 5 | 605 | 97 | |
| GenBank mRNA | BC135814.1 | - | 4 - 5 | 605 | 97 | |
More Information
Set Membership: |
|

g6pc2
Genomic Map