Assay Details
| Gene Symbol: cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204020.4 | NP_989351.3 | 4 - 5 | 415 | 57 | |
| XM_012970268.1 | XP_012825722.1 | 4 - 5 | 447 | 57 | ||
| GenBank mRNA | BC064226.1 | - | 4 - 5 | 399 | 57 | |
| BC075535.1 | - | 4 - 5 | 413 | 57 | ||
| CR848140.2 | - | 4 - 5 | 415 | 57 | ||
More Information
Set Membership: |
|

cd2bp2
Genomic Map