Assay Details
| Gene Symbol: mrpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016279.2 | NP_001016279.1 | 1 | 157 | 58 | |
| XM_012969761.2 | XP_012825215.1 | 2 | 235 | 58 | ||
| XM_012969762.2 | XP_012825216.1 | 2 | 260 | 58 | ||
| GenBank mRNA | BC157287.1 | - | 1 | 157 | 58 | |
| CR761612.1 | - | 1 | 542 | 58 | ||
More Information
Set Membership: |
|

mrpl41
Genomic Map