Assay Details
| Gene Symbol: trmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016423.3 | NP_001016423.2 | 3 - 4 | 452 | 89 | |
| XM_012962292.2 | XP_012817746.1 | 3 - 4 | 582 | 89 | ||
| XM_018093244.1 | XP_017948733.1 | 3 - 4 | 583 | 89 | ||
| GenBank mRNA | BC158168.1 | - | 3 - 4 | 452 | 89 | |
| BC171030.1 | - | 2 - 3 | 404 | 89 | ||
More Information
Set Membership: |
|

trmo
Genomic Map