Assay Details
| Gene Symbol: trmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016423.3 | NP_001016423.2 | 5 - 6 | 1193 | 75 | |
| XM_012962292.2 | XP_012817746.1 | 5 - 6 | 1323 | 75 | ||
| XM_018093244.1 | XP_017948733.1 | 5 - 6 | 1249 | 75 | ||
| GenBank mRNA | BC158168.1 | - | 5 - 6 | 1193 | 75 | |
| BC171030.1 | - | 4 - 5 | 1145 | 75 | ||
More Information
Set Membership: |
|

trmo
Genomic Map