Assay Details
Gene Symbol: pofut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078711.1 | NP_001072179.1 | 5 - 6 | 727 | 72 | |
XM_012970523.2 | XP_012825977.1 | 4 - 5 | 612 | 72 | ||
XM_018097146.1 | XP_017952635.1 | 5 - 6 | 2938 | 72 | ||
GenBank mRNA | DQ139950.1 | - | 5 - 6 | 727 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
