Assay Details
| Gene Symbol: ric3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078773.1 | NP_001072241.1 | 3 - 4 | 420 | 63 | |
| GenBank mRNA | BC118843.1 | - | 3 - 4 | 420 | 63 | |
More Information
Set Membership: |
|

ric3
Genomic Map