Assay Details
| Gene Symbol: plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103075.1 | NP_001096545.1 | 12 - 13 | 1719 | 102 | |
| GenBank mRNA | BC136055.1 | - | 12 - 13 | 1719 | 102 | |
More Information
Set Membership: |
|

plk2
Genomic Map