Assay Details
| Gene Symbol: brf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103079.1 | NP_001096549.1 | 2 - 3 | 245 | 63 | |
| GenBank mRNA | BC136234.1 | - | 2 - 3 | 245 | 63 | |
| CR942474.2 | - | 2 - 3 | 257 | 63 | ||
More Information
Set Membership: |
|

brf2
Genomic Map