Assay Details
| Gene Symbol: hdgfl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103080.1 | NP_001096550.1 | 4 - 5 | 500 | 89 | |
| XM_012958473.2 | XP_012813927.1 | 4 - 5 | 901 | 89 | ||
| GenBank mRNA | BC136239.1 | - | 4 - 5 | 500 | 89 | |
More Information
Set Membership: |
|

hdgfl3
Genomic Map