Assay Details
| Gene Symbol: hdgfl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103080.1 | NP_001096550.1 | 5 - 6 | 647 | 76 | |
| GenBank mRNA | BC136239.1 | - | 5 - 6 | 647 | 76 | |
More Information
Set Membership: |
|

hdgfl3
Genomic Map