Assay Details
| Gene Symbol: cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105514.1 | NP_001098984.1 | 2 - 3 | 431 | 79 | |
| GenBank mRNA | CT030066.2 | - | 2 - 3 | 514 | 79 | |
| CU025129.1 | - | 2 - 3 | 431 | 79 | ||
More Information
Set Membership: |
|

cyr61
Genomic Map