Assay Details
| Gene Symbol: cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105514.1 | NP_001098984.1 | 4 - 5 | 990 | 87 | |
| GenBank mRNA | CU025129.1 | - | 4 - 5 | 990 | 87 | |
More Information
Set Membership: |
|

cyr61
Genomic Map