Assay Details
| Gene Symbol: psmb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110042.1 | NP_001103512.1 | 1 - 2 | 350 | 57 | |
| XM_012954239.1 | XP_012809693.1 | 1 - 2 | 428 | 57 | ||
| GenBank mRNA | BC153713.1 | - | 1 - 2 | 350 | 57 | |
More Information
Set Membership: |
|

psmb8
Genomic Map