Assay Details
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110050.1 | NP_001103520.2 | 7 - 8 | 1007 | 66 | |
XM_012956327.2 | XP_012811781.1 | 8 - 9 | 1819 | 66 | ||
XM_012956331.2 | XP_012811785.1 | 8 - 9 | 1869 | 66 | ||
XM_012956333.2 | XP_012811787.1 | 8 - 9 | 1816 | 66 | ||
XM_012956334.2 | XP_012811788.1 | 8 - 9 | 1865 | 66 | ||
XM_012956335.2 | XP_012811789.1 | 8 - 9 | 1805 | 66 | ||
XM_018091054.1 | XP_017946543.1 | 8 - 9 | 1820 | 66 | ||
GenBank mRNA | BC153338.1 | - | 7 - 8 | 1007 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
