Assay Details
| Gene Symbol: trem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110052.1 | NP_001103522.1 | 2 - 3 | 458 | 103 | |
| XM_012967981.2 | XP_012823435.1 | 2 - 3 | 524 | 103 | ||
| XM_012968030.1 | XP_012823484.1 | 2 - 3 | 479 | 103 | ||
| GenBank mRNA | BC153718.1 | - | 2 - 3 | 458 | 103 | |
More Information
Set Membership: |
|

trem2
Genomic Map