Assay Details
| Gene Symbol: rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112943.1 | NP_001106414.1 | 5 - 6 | 1718 | 59 | |
| XM_012953956.2 | XP_012809410.1 | 6 - 7 | 1945 | 59 | ||
| XM_012953957.2 | XP_012809411.1 | 6 - 7 | 1980 | 59 | ||
| XM_012953958.2 | XP_012809412.1 | 7 - 8 | 2062 | 59 | ||
| XM_012953959.2 | XP_012809413.1 | 7 - 8 | 2097 | 59 | ||
| GenBank mRNA | BC118910.1 | - | 5 - 6 | 1719 | 59 | |
| BC153348.1 | - | 5 - 6 | 1718 | 59 | ||
More Information
Set Membership: |
|

rab11fip2
Genomic Map