Assay Details
| Gene Symbol: cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112946.1 | NP_001106417.1 | 4 - 5 | 1013 | 72 | |
| GenBank mRNA | BC153352.1 | - | 4 - 5 | 1013 | 72 | |
More Information
Set Membership: |
|

cpe
Genomic Map