Assay Details
| Gene Symbol: pmm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112950.1 | NP_001106421.1 | 3 - 4 | 289 | 75 | |
| GenBank mRNA | BC153361.1 | - | 3 - 4 | 289 | 75 | |
More Information
Set Membership: |
|

pmm1
Genomic Map