Assay Details
| Gene Symbol: pigs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112953.1 | NP_001106424.1 | 4 - 5 | 378 | 59 | |
| XM_012956342.2 | XP_012811796.1 | 4 - 5 | 524 | 59 | ||
| GenBank mRNA | BC153365.1 | - | 4 - 5 | 378 | 59 | |
More Information
Set Membership: |
|

pigs
Genomic Map