Assay Details
| Gene Symbol: jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112955.1 | NP_001106426.1 | 10 - 11 | 2569 | 60 | |
| GenBank mRNA | BC153370.1 | - | 10 - 11 | 2569 | 60 | |
More Information
Set Membership: |
|

jmy
Genomic Map