Assay Details
| Gene Symbol: fam207a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112959.1 | NP_001106430.1 | 4 - 5 | 495 | 74 | |
| XM_018097030.1 | XP_017952519.1 | 3 - 4 | 296 | 74 | ||
| GenBank mRNA | BC153710.1 | - | 4 - 5 | 495 | 74 | |
| BC158169.1 | - | 4 - 5 | 497 | 74 | ||
| CR761930.2 | - | 4 - 5 | 494 | 74 | ||
More Information
Set Membership: |
|

fam207a
Genomic Map