Assay Details
| Gene Symbol: lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112972.1 | NP_001106443.1 | 5 - 6 | 418 | 69 | |
| GenBank mRNA | BC153732.1 | - | 5 - 6 | 418 | 69 | |
More Information
Set Membership: |
|

lrrc34
Genomic Map