Assay Details
| Gene Symbol: tcp11l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112989.1 | NP_001106460.1 | 9 - 10 | 1532 | 71 | |
| XM_012960517.1 | XP_012815971.1 | 9 - 10 | 1371 | 71 | ||
| XM_012960518.2 | XP_012815972.1 | 9 - 10 | 1643 | 71 | ||
| GenBank mRNA | BC154074.1 | - | 9 - 10 | 1532 | 71 | |
More Information
Set Membership: |
|

tcp11l1
Genomic Map