Assay Details
| Gene Symbol: dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113004.1 | NP_001106475.1 | 10 - 11 | 1480 | 57 | |
| XM_012960526.2 | XP_012815980.1 | 11 - 12 | 1583 | 57 | ||
| GenBank mRNA | BC154097.1 | - | 10 - 11 | 1480 | 57 | |
More Information
Set Membership: |
|

dalrd3
Genomic Map