Assay Details
| Gene Symbol: tacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113018.1 | NP_001106489.1 | 4 - 5 | 1333 | 66 | |
| GenBank mRNA | BC154685.1 | - | 4 - 5 | 1333 | 66 | |
More Information
Set Membership: |
|

tacr1
Genomic Map