Assay Details
| Gene Symbol: lypd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113032.3 | NP_001106503.1 | 3 - 4 | 427 | 80 | |
| GenBank mRNA | BC154706.1 | - | 625 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lypd6| Gene Symbol: lypd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113032.3 | NP_001106503.1 | 3 - 4 | 427 | 80 | |
| GenBank mRNA | BC154706.1 | - | 625 | 80 | ||
Set Membership: |
|