Assay Details
| Gene Symbol: tmprss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113037.1 | NP_001106508.1 | 2 - 3 | 180 | 69 | |
| XM_012956358.2 | XP_012811812.1 | 1 - 2 | 725 | 69 | ||
| GenBank mRNA | BC154712.1 | - | 2 - 3 | 180 | 69 | |
More Information
Set Membership: |
|

tmprss12
Genomic Map