Assay Details
| Gene Symbol: tmprss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113037.1 | NP_001106508.1 | 5 - 6 | 784 | 79 | |
| XM_012956358.2 | XP_012811812.1 | 4 - 5 | 1329 | 79 | ||
| GenBank mRNA | BC154712.1 | - | 5 - 6 | 784 | 79 | |
More Information
Set Membership: |
|

tmprss12
Genomic Map