Assay Details
| Gene Symbol: capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113040.1 | NP_001106511.1 | 3 - 4 | 1582 | 58 | |
| GenBank mRNA | BC154716.1 | - | 3 - 4 | 1582 | 58 | |
More Information
Set Membership: |
|

capn15
Genomic Map