Assay Details
| Gene Symbol: ndufb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113058.1 | NP_001106529.1 | 3 - 4 | 323 | 63 | |
| GenBank mRNA | BC154898.1 | - | 3 - 4 | 323 | 63 | |
More Information
Set Membership: |
|

ndufb2
Genomic Map