Assay Details
| Gene Symbol: adora1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113069.1 | NP_001106540.1 | 2 - 3 | 566 | 70 | |
| GenBank mRNA | BC154912.1 | - | 2 - 3 | 566 | 70 | |
More Information
Set Membership: |
|

adora1
Genomic Map