Assay Details
| Gene Symbol: ciao3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113086.1 | NP_001106557.1 | 6 - 7 | 728 | 73 | |
| GenBank mRNA | BC090375.1 | - | 5 - 6 | 994 | 73 | |
| BC154937.1 | - | 6 - 7 | 728 | 73 | ||
More Information
Set Membership: |
|

ciao3
Genomic Map