Assay Details
| Gene Symbol: g6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113088.1 | NP_001106559.1 | 3 - 4 | 461 | 97 | |
| GenBank mRNA | BC154943.1 | - | 3 - 4 | 461 | 97 | |
More Information
Set Membership: |
|

g6pc
Genomic Map