Assay Details
Gene Symbol: mcrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113097.1 | NP_001106568.1 | 3 - 4 | 178 | 84 | |
XM_012965941.2 | XP_012821395.1 | 3 - 4 | 264 | 84 | ||
XM_012965942.2 | XP_012821396.1 | 3 - 4 | 265 | 84 | ||
XM_012965943.2 | XP_012821397.1 | 4 - 5 | 291 | 84 | ||
XM_012965944.2 | XP_012821398.1 | 3 - 4 | 233 | 84 | ||
XM_012965945.2 | XP_012821399.1 | 4 - 5 | 273 | 84 | ||
GenBank mRNA | BC155036.1 | - | 3 - 4 | 178 | 84 | |
BC161072.1 | - | 3 - 4 | 181 | 84 | ||
CR762021.2 | - | 3 - 4 | 174 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
