Assay Details
| Gene Symbol: prkab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113101.1 | NP_001106572.1 | 7 - 8 | 858 | 73 | |
| GenBank mRNA | BC155097.1 | - | 7 - 8 | 858 | 73 | |
More Information
Set Membership: |
|

prkab2
Genomic Map