Assay Details
| Gene Symbol: btnl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113107.2 | NP_001106578.2 | 5 - 6 | 969 | 83 | |
| GenBank mRNA | BC155397.1 | - | 5 - 6 | 942 | 83 | |
More Information
Set Membership: |
|

btnl8
Genomic Map