Assay Details
| Gene Symbol: btnl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113107.2 | NP_001106578.2 | 6 - 7 | 1065 | 103 | |
| GenBank mRNA | BC155397.1 | - | 6 - 7 | 1038 | 103 | |
More Information
Set Membership: |
|

btnl8
Genomic Map