Assay Details
| Gene Symbol: renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113110.1 | NP_001106581.1 | 2 - 3 | 225 | 80 | |
| XM_018096131.1 | XP_017951620.1 | 2 - 3 | 170 | 80 | ||
| GenBank mRNA | BC155401.1 | - | 2 - 3 | 225 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018096131.1 transcripts.
Set Membership: |
|

renbp
Genomic Map