Assay Details
| Gene Symbol: pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113147.1 | NP_001106618.1 | 1 - 2 | 447 | 78 | |
| XM_012964237.1 | XP_012819691.1 | 2 - 3 | 562 | 78 | ||
| XM_012964238.1 | XP_012819692.1 | 2 - 3 | 530 | 78 | ||
| XM_012964240.2 | XP_012819694.1 | 2 - 3 | 579 | 78 | ||
| GenBank mRNA | BC155461.1 | - | 1 - 2 | 447 | 78 | |
More Information
Set Membership: |
|

pter
Genomic Map