Assay Details
Gene Symbol: tnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113671.1 | NP_001107143.1 | 2 - 3 | 283 | 97 | |
GenBank mRNA | AM041993.1 | - | 2 - 3 | 283 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
